Gas washing bottles, with filter candle, VitraPOR®
Toimittaja: ROBU GLASFILTER
42093 42090 42092 42094 42130 42131 42132 42133 42134 42091
ROBU42093EA 92.8 EUR
ROBU42093 ROBU42090 ROBU42092 ROBU42094 ROBU42130 ROBU42131 ROBU42132 ROBU42133 ROBU42134 ROBU42091
Gas washing bottles, with filter candle, VitraPOR®
Pullot Kaasunpesupullot
They are made of borosilicate glass 3.3 and the versatile and recyclable material offers good properties for the most diverse fields.
  • Resistant to chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
  • With screw cap and silicone washer