Chromatographic columns with fused filter discs, VitraPOR®
Toimittaja: ROBU GLASFILTER
31202 31101 31102 31103 31201 31203 31204 31104
ROBU31202EA 304 EUR
ROBU31202 ROBU31101 ROBU31102 ROBU31103 ROBU31201 ROBU31203 ROBU31204 ROBU31104
Chromatographic columns with fused filter discs, VitraPOR®
Kromatografiakolonnit
VitraPOR® chromatography columns are made of borosilicate glass 3.3. The versatile and recyclable material makes it suitable for the most diverse fields.
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Glass filters are divided into porosity classes ranging from 1,0 to 500 µm
Toimitustiedot: Filter tubes are available in various diameters and pore sizes.