Micro-Immersion Filters
Toimittaja: ROBU GLASFILTER
25100 25101 25102 25103 25104
ROBU25100EA 130 EUR
ROBU25100 ROBU25101 ROBU25102 ROBU25103 ROBU25104
Micro-Immersion Filters
Suodattimet Suodatinputket
ROBU® micro-immersion filter with VitraPOR® glass filter are designed for efficient reverse filtration of liquids. Ideal for applications requiring fine particle separation directly within the vessel.
  • For reverse filtration
  • Integrated VitraPOR® filter
  • Reusable, chemical resistant
  • Suitable for laboratory and process use
  • Various porosities available
  • Easy immersion into vessels
ROBU immersion filters equipped with a VitraPOR glass filter enable highly efficient reverse filtration directly within containers or vessels. Designed for laboratory and