Immersion filters, VitraPOR®
Toimittaja: ROBU GLASFILTER
24202 24201 24203 24204 24350 24351 24352 24353 24354 24200
511-2312EA 39.3 EUR
511-2312 511-2311 511-2313 511-2314 ROBU24350 ROBU24351 ROBU24352 ROBU24353 ROBU24354 511-2310
Immersion filters, VitraPOR®
Suodattimet Suodatinputket
These immersion filters are suitable for reverse filtration.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of borosilicate glass 3.3.