ROBU® Rubber Sleeves
Toimittaja: ROBU GLASFILTER
20701 20702 20703 20704
ROBU20701EA 13.2 EUR
ROBU20701 ROBU20702 ROBU20703 ROBU20704
ROBU® Rubber Sleeves
Pullotuet Tukihylsyt
ROBU® Rubber Sleeves for Crucibles provide secure sealing for filter crucibles and corresponding adapters. Made from high-quality rubber, they ensure a tight fit and prevent leaks during filtration processes, offering durability and reliability in laboratory applications.
  • Secure sealing for filter crucibles
  • Prevents leaks during filtration
  • High-quality rubber construction
  • Durable and long-lasting
  • Ensures tight fit with corresponding adapters
  • Resistant to chemicals and high temperatures
  • Easy to install and replace
The ROBU Rubber sleeves for crucibles