Vacuum-/gas distribution filters, VitraPOR®
Toimittaja: ROBU GLASFILTER
18500 18501 18502 18503 18504
ROBU18500EA 48.1 EUR
ROBU18500 ROBU18501 ROBU18502 ROBU18503 ROBU18504
Vacuum-/gas distribution filters, VitraPOR®
Suodattimet Nesteen prosessisuodattimet
These filters are suitable for degassing and filtration of solvents e.g. in chromatography.
  • Good chemical resistance
  • Good temperature resistance
Made of borosilicate glass.