Micro-Filter Candles
Toimittaja: ROBU GLASFILTER
18100 18123 18101 18102 18104 18105 18110 18111 18112 18113 18114 18115 18120 18122 18124 18125 18130 18131 18132 18133 18134 18140 18141 18142 18143 18144 18121 18103
ROBU18100EA 245 EUR
ROBU18100 ROBU18123 ROBU18101 ROBU18102 ROBU18104 ROBU18105 ROBU18110 ROBU18111 ROBU18112 ROBU18113 ROBU18114 ROBU18115 ROBU18120 ROBU18122 ROBU18124 ROBU18125 ROBU18130 ROBU18131 ROBU18132 ROBU18133 ROBU18134 ROBU18140 ROBU18141 ROBU18142 ROBU18143 ROBU18144 ROBU18121 ROBU18103
Micro-Filter Candles
Suodattimet Suodatinputket
VitraPOR® micro-filter candles provide precise filtration in compact setups. Ideal for low-volume applications requiring chemical resistance and fine particle retention.
  • VitraPOR® porous glass filter
  • Fine and uniform pore structure
  • Compact size
  • High chemical and thermal resistance
  • Suitable for gases and liquids
  • Reusable
  • Ideal for laboratory and process applications
ROBU Micro-Filter Candles with integrated VitraPOR filter are high-precision filtration elements designed for small-volume and confined-space applications. Made entirely of borosilicate