Filter discs, biplane, series 16, porosity 2, VitraPOR®
Toimittaja: ROBU GLASFILTER
16302 16402 16502 16802 16602 16702 16902 16112 16122 16102 16202 16252 62002 61902 62102 61302 62802 62702 62602 62502 62402 63502 63802 63002 62902 61702 61602 61502 61402 62302 62202 61802
ROBU16302EA 93.3 EUR
ROBU16302 ROBU16402 ROBU16502 ROBU16802 ROBU16602 ROBU16702 ROBU16902 ROBU16112 ROBU16122 ROBU16102 ROBU16202 ROBU16252 ROBU62002 ROBU61902 ROBU62102 ROBU61302 ROBU62802 ROBU62702 ROBU62602 ROBU62502 ROBU62402 ROBU63502 ROBU63802 ROBU63002 ROBU62902 ROBU61702 ROBU61602 ROBU61502 ROBU61402 ROBU62302 ROBU62202 ROBU61802
Filter discs, biplane, series 16, porosity 2, VitraPOR®
Suodattimet Suodatinkiekot
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.