Foot adapter
Toimittaja: R L ENTERPRISES
2119
RLAA2119EA 32.4 EUR
RLAA2119
Foot adapter
Kourat Kaksoismuhvit
Adapter manufactured out of aluminium with boss to fit scaffold feet.
Accepts three 13 mm diameter rods or tubes at right angles (2 perpendicular to the boss and the third in line with the boss), with alien screws for fixing. One allen key provided per pack of ten.