Jerricans, Narrow Neck, HDPE, UN-X Approved
Toimittaja: RIXIUS
6-0300-200-61 6030005051WC 6030010051WC
RIXI6-0300-200-61EA 1200 EUR
RIXI6-0300-200-61 RIXI6030005051WC RIXI6030010051WC
Jerricans, Narrow Neck, HDPE, UN-X Approved
Pullot Jerry-kannut
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
  • UN canister with X-approval
Universal canister for industry, trade and laboratories. Ideal for packaging, storage and transportation.