Jerricans, Narrow Neck, HDPE
Toimittaja: RIXIUS
6-0200-200-61 6-0200-250-61 6020002545WC 6020003045WC 6020020061WC 6020025061WC 6020030061WC 6020005045WC 6020002045WC 60200050452WC
216-5334EA 14.9 EUR
216-5334 216-5376 RIXI6020002545WC RIXI6020003045WC RIXI6020020061WC RIXI6020025061WC RIXI6020030061WC RIXI6020005045WC RIXI6020002045WC RIXI60200050452WC
Jerricans, Narrow Neck, HDPE
Pullot Jerry-kannut
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
  • UN canister with Y approval
Universal canister for industry, trade, and laboratories. Ideal for packaging, storage and transportation. With UN or Y approval.
Tilaustiedot: Canisters are supplied with or without screw cap. Additional compatible screw cap types are available separately.