Bung barrels
Toimittaja: RIXIUS
5-2203-220-MA
RIXI5-2203-220-MAEA 168 EUR
RIXI5-2203-220-MA
Bung barrels
Tynnyrit, tynnyrit, rummut
HDPE, blue.
  • Good chemical resistance
  • Stable and robust
Suitable for storage, disposal and transport of laboratory materials. With S70×6 and S56×4 plug, PE gasket and two sealed caps.
Sertifikaatit: UN approval: 1H1/X/250