Jars, Wide Mouth, Round, Rixius
Toimittaja: RIXIUS
4-0100-0500-70 4-0100-1000-80 4-0100-0250-60 4-0100-0500-80 4-0100-0750-80 4-0100-0100-4 40100100080OD 40100100080WC
216-4010EA 2.1 EUR
216-4010 RIXI4-0100-1000-80 RIXI4-0100-0250-60 RIXI4-0100-0500-80 RIXI4-0100-0750-80 RIXI4-0100-0100-4 RIXI40100100080OD RIXI40100100080WC
Jars, Wide Mouth, Round, Rixius
Astiat ja purkit Astiat, yleiskäyttö
Jars, wide mouth, made of HDPE, without or with screw lid.
  • Good chemical resistance
  • Good temperature resistance
  • Lid must be ordered seperately