Chemical bottles, wide mouth, square
Toimittaja: RIXIUS
2-0600-0950-70 2-0604-0250-50 2-0600-0500-64 2-0600-0250-50
RIXI2-0600-0950-70EA 227 EUR
RIXI2-0600-0950-70 RIXI2-0604-0250-50 RIXI2-0600-0500-64 RIXI2-0600-0250-50
Chemical bottles, wide mouth, square
Pullot Muovipullot
HDPE.
  • Good chemical resistance
  • Good temperature resistance
Tilaustiedot: Screw caps must be ordered separately.