Test sieves with perforated plates
Toimittaja: Retsch GmbH
60.198.003150 60.198.004000 60.198.006300 60.198.008000 60.198.010000 60.162.045000 60.162.008000 60.162.016000 60.162.063000 60.162.100000 60.162.003150 60.138.011200 60.198.012500 60.198.013200 60.154.005600 60.154.008000 60.198.001000 60.198.002000 60.198.004500 60.198.005000 60.146.002000 60.146.003150 60.146.003550 60.146.004000 60.146.004500 60.146.005000 60.146.005600 60.146.002500 60.190.004000 60.190.008000 60.190.016000 60.198.002800 60.198.003350 60.198.002240 60.190.005600 60.190.011200 60.190.022400 60.190.031500 60.198.007100 60.198.025000 60.198.016000 60.198.045000 60.162.005000 60.198.020000 60.138.004000 60.138.008000 60.198.011200 60.198.031500 60.162.006300 60.162.012500 60.162.025000 60.162.050000 60.162.080000 60.154.020000 60.154.022400 60.154.031500 60.154.040000 60.154.045000 60.154.056000 60.154.063000 60.154.080000 60.154.090000 60.154.125000 60.154.005000 60.154.006300 60.154.011200 60.154.010000 60.154.012500 60.154.014000 60.154.016000 60.146.010000 60.146.011200 60.146.014000 60.146.020000 60.146.022400 60.146.040000 60.146.045000 60.138.010000 60.138.037500 60.198.003550 60.198.001600 60.198.002500 60.198.005600 60.198.004750 60.162.002000 60.162.010000 60.162.014000 60.162.020000 60.162.001000 60.198.063000
RETS60.198.003150EA 295 EUR
RETS60.198.003150 RETS60.198.004000 RETS60.198.006300 RETS60.198.008000 RETS60.198.010000 RETS60.162.045000 RETS60.162.008000 RETS60.162.016000 RETS60.162.063000 RETS60.162.100000 RETS60.162.003150 RETS60.138.011200 RETS60.198.012500 RETS60.198.013200 RETS60.154.005600 RETS60.154.008000 RETS60.198.001000 RETS60.198.002000 RETS60.198.004500 RETS60.198.005000 RETS60.146.002000 RETS60.146.003150 RETS60.146.003550 RETS60.146.004000 RETS60.146.004500 RETS60.146.005000 RETS60.146.005600 RETS60.146.002500 RETS60.190.004000 RETS60.190.008000 RETS60.190.016000 RETS60.198.002800 RETS60.198.003350 RETS60.198.002240 RETS60.190.005600 RETS60.190.011200 RETS60.190.022400 RETS60.190.031500 RETS60.198.007100 RETS60.198.025000 RETS60.198.016000 RETS60.198.045000 RETS60.162.005000 RETS60.198.020000 RETS60.138.004000 RETS60.138.008000 RETS60.198.011200 RETS60.198.031500 RETS60.162.006300 RETS60.162.012500 RETS60.162.025000 RETS60.162.050000 RETS60.162.080000 RETS60.154.020000 RETS60.154.022400 RETS60.154.031500 RETS60.154.040000 RETS60.154.045000 RETS60.154.056000 RETS60.154.063000 RETS60.154.080000 RETS60.154.090000 RETS60.154.125000 RETS60.154.005000 RETS60.154.006300 RETS60.154.011200 RETS60.154.010000 RETS60.154.012500 RETS60.154.014000 RETS60.154.016000 RETS60.146.010000 RETS60.146.011200 RETS60.146.014000 RETS60.146.020000 RETS60.146.022400 RETS60.146.040000 RETS60.146.045000 RETS60.138.010000 RETS60.138.037500 RETS60.198.003550 RETS60.198.001600 RETS60.198.002500 RETS60.198.005600 RETS60.198.004750 RETS60.162.002000 RETS60.162.010000 RETS60.162.014000 RETS60.162.020000 RETS60.162.001000 RETS60.198.063000
Test sieves with perforated plates
Seulat Testisiivilät
Sieves consist of a solid stainless steel sieve frame of high stability for reliable sieving results.
  • High degree of corrosion resistance
  • Maximum stability and optimum sealing when used in sieve stacks
The individual laser engraving of each test sieve provides a clear and accurate labelling with full traceability. Clear and precise labelling of the sieves with full traceability based on individualized laser engraving. The sieve fabric is precisely joined into the frame and tautened.