Carbon standards
Toimittaja: REAGECON
TOIC10 TOIC100 TOIC1M TOC50 TIC1M TOC1M TIC500 TOC500 TOC2M TOC5 TIC5 TOC100 TOC10 TOC20 TOC15 TOC250
REGETOIC10EA 600 EUR
REGETOIC10 REGETOIC100 701-0049 REGETOC50 REGETIC1M REGETOC1M REGETIC500 REGETOC500 REGETOC2M REGETOC5 REGETIC5 REGETOC100 REGETOC10 REGETOC20 REGETOC15 REGETOC250
Carbon standards
Standardiaineet ja vertailumateriaalit Cationic Standards Carbon Standards
These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.
These standards are independent, traceable and certified.
Sertifikaatit: Accredited to IEC/ISO 17025 and ISO 17034.