System Suitability Set to USP for Total Organic Carbon (TOC) Suitable for Use with Anatel A643 Analyser
Toimittaja: REAGECON
ISTOC1265
REGEISTOC1265EA 943 EUR
REGEISTOC1265
System Suitability Set to USP for Total Organic Carbon (TOC) Suitable for Use with Anatel A643 Analyser
Standardiaineet ja vertailumateriaalit Ympäristöanalytiikan standardit ja vertailumateriaalit
Reagecon's system suitability set to from USP for Total Organic Carbon (TOC) is a product of extremely high specification and purity. The product is produced from ultra pure raw materials including water that is prepared by a special proprietary process. The product is presented in bottles that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.