Turbidity standards, mid range 200 to 1000 NTU
Toimittaja: REAGECON
CRS-200-100 CRS-200-500 CRS-400-100 CRS-400-500 CRS-500-100 CRS-500-500 CRSR-1000-500 CRSR-400-500 CRSR-500-500 CRSR-1000-100 CRSR-400-100 CRSR-500-100
REGECRS-200-100EA 475 EUR
REGECRS-200-100 REGECRS-200-500 REGECRS-400-100 REGECRS-400-500 REGECRS-500-100 REGECRS-500-500 REGECRSR-1000-500 REGECRSR-400-500 REGECRSR-500-500 REGECRSR-1000-100 REGECRSR-400-100 REGECRSR-500-100
Turbidity standards, mid range 200 to 1000 NTU
Standardiaineet ja vertailumateriaalit Sameustandardit
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
  • Non toxic and non carcinogenic microspheres
  • Highly accurate with well defined spherical shape
  • Traceable to NIST
  • US EPA approved
  • Ready to use- particles stay in suspension