Etikkahappo 0.1 mol/l (0.1 N) volumetric solution

Toimittaja: REAGECON

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

CH20101
REGECH20101EA 638 EUR
REGECH20101
Etikkahappo 0.1 mol/l (0.1 N) volumetric solution
Etikkahappo
Kaava: H₃CCOOH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR