Arsenic standard solution, for limit tests
Toimittaja: REAGECON
5000500C
REGE5000500CEA 219 EUR
REGE5000500C
Arsenic standard solution, for limit tests
Standardiaineet ja vertailumateriaalit Cationic Standards Arsenic Standards
Concentrate to make arsenic standard solution (10 ppm as As) as outlined in Chapter 4 (4.1.2) of Ph. Eur.