Agar

Toimittaja: G Bioscienes

RC-004 RC-003
RC-004EA 786 EUR
RC-004 RC-003
Agar
Agar
Sulamispiste: 90 °C
MDL numero: MFCD00081288
CAS-numero: 9002-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR