Deep well blocks, 96-well, low profile
Toimittaja: RATIOLAB
8101012
RATI8101012EA 201 EUR
RATI8101012
Deep well blocks, 96-well, low profile
Kuoppalevyt Syväkuoppalevyt
These 1.2 ml low-profile deep-well plates made from clear PP are ideal for pharmaceutics, biotechnology and diagnostics.
  • Well with significantly larger cross section and raised rims for reduced cross-contamination
  • Autoclavable at 121 °C, 20 minutes
  • Temperature resistant to –80 °C
  • 8 x 12 wells with alphanumeric code in micro test plate format
  • Precise design, for use in automatic pipetting systems for High-Throughput-Screening (HTS)
  • Optimum space for bar code labels on the side walls
  • Space-saving and stackable
Being of the same standard dimensions (More Product Information