Cryoboxes
Toimittaja: RATIOLAB
5150021 5150022 5150023 5150002 5150003 5150012 5150013 5150014 5150011
RATI5150021EA 71.4 EUR
RATI5150021 RATI5150022 RATI5150023 RATI5150002 RATI5150003 RATI5150012 RATI5150013 RATI5150014 RATI5150011
Cryoboxes
Laatikot Kryogeenipullolaatikot
PC, coloured, with clear lid.
  • Temperature-resistant from −190 to 121 °C
  • Drain bores at the bottom for dew liquids
  • By stacking corners the boxes are suitable for safe transport
With slip lid and variable grid insert.