RNA isolation, total RNA kit II, E.Z.N.A.®
Toimittaja: OMEGA BIO-TEK
R6934-01 R6934-00 R6934-02
R6934-01EA 262 EUR
R6934-01 R6934-00 R6934-02
RNA isolation, total RNA kit II, E.Z.N.A.®
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Total RNA kit II is designed for isolating total cellular RNA from tissues rich fibrous and fatty tissues such as skeletal muscle, heart, brain and adipose tissues. Compared to other standard silica-column procedures, the E.Z.N.A.® Total RNA kit II provides higher yield and better quality of RNA from all types of tissue. This kit combines phenol/guanidine-base lysis and the silica membrane purification of RNA technology to provide a rapid and easy method of the isolation of total RNA from any tissue sample. RNA purified using the E.Z.N.A.® Total RNA method is ready for applications such as RT-PCR, Northern blotting, poly A+ RNA (mRNA) purification, nuclease protection, and in vitro translation.
  • High quality RNA in 40 minutes
  • Optimised buffers ensure the reproducible results
  • Suited for all types of biological samples
  • Purified RNA suitable for most downstream applications