Vesi nukleaasiton, Fermantas

Toimittaja: Thermo Fisher Scientific

R0581 R0582
R0581EA 31.17 EUR
R0581 R0582
Vesi nukleaasiton, Fermantas
Vesi

This nuclease-free water is deionized and 0.22 µM membrane-filtered nuclease-free water


  • Free of endo, exodeoxyribonucleases, ribonucleases, and phosphatases


It is ideal for all molecular biology applications.

Kaava: H₂O
Kiehumispiste: 100 °C (1013 hPa)
Sulamispiste: 0 °C
Tiheys: 1 g/cm³ (4 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011332
CAS-numero: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR