Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Toimittaja: VWR Collection
QNTS081-3PTWH0XLIDEA 0 EUR
QNTS081-3PTWH0XLID
Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Työvaatteet
These coats have both zipper and pockets.
  • Hanger in collar
  • Stitched collar patch with labe
  • Antistatic knitted cuffs
  • One breast pocket and two hip pockets
  • Two way zip in front
The coats contain press studs on left breast for PES clipband and also lap seams for tight and secure seam-enclosure.