Multi layer adhesive floor mats
Toimittaja: PURUS
PM264538W-HT PM264538G PM264538W PM184538W PM184538W-HT PM364534B PM364564G PM264538B-HT PM243638W PM264538B
PURUPM264538W-HTEA 337 EUR
PURUPM264538W-HT PURUPM264538G PURUPM264538W PURUPM184538W PURUPM184538W-HT PURUPM364534B PURUPM364564G PURUPM264538B-HT PURUPM243638W PURUPM264538B
Multi layer adhesive floor mats
Matot Puhdastilamatot
A choice of either 30 or 60 layer adhesive entrance mats are manufactured from low density polyethylene. The mats control contamination by removing debris from the soles of shoes and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.
  • Antimicrobial agent protects against the growth of bacteria and fungi
  • Non-slip, low profile, individually numbered layers
  • Reduced maintenance and cleaning costs
Colour: Clear, white, grey or blue