Safety spectacles, Pan View, PW38
Toimittaja: Portwest
PW38BKR PW38CLR PW38COR PW38SOR
PRTWPW38BKREA 3.6 EUR
PRTWPW38BKR PRTWPW38CLR PRTWPW38COR PRTWPW38SOR
Safety spectacles, Pan View, PW38
Suojalasit
These sleek and sporty style glasses are designed for safety and comfort. The Pan View features lightweight construction and soft touch temples. The single wrap-around lens offers protection from the sides and front.
  • 99% UV protection
  • One piece curved lens with moulded nose-bridge
  • Soft arm tips and rubber nose-bridge for ultra comfort
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
  • Retail bag which aids presentation for retail sales
  • Free spec cord
Sertifikaatit: EN 166 1 FT; EN 170 2C-1.2 (CLEAR); EN 172More Product Information