Gas filters, P906
Toimittaja: Portwest
P906BKR
PRTWP906BKREA 92.1 EUR
PRTWP906BKR
Gas filters, P906
Suodattimet Kaasusuodattimet
P906 filters are anti-gas filter that protects against contaminant gases. Equipped with a standard threaded connection conforming to EN 148-1 and can be used on full face masks equipped with the same standard connection.
  • Lightweight and comfortable
  • Individually packed for vending machines
These gas filter protects from organic gases and vapours where the boiling point is higher than 65 °C. Classified as a class 2 filter, they can be used to protect against harmful substances, solvents and gases.
Sertifikaatit: EN 148 -1, EN 14387 Type A Class 2