Disposable particle filtering respirators, P301, BizTex®
Toimittaja: Portwest
P301WHR
PRTWP301WHREA 55.2 EUR
PRTWP301WHR
Disposable particle filtering respirators, P301, BizTex®
Hengityssuojaimet Kertakäyttöiset hengityssuojaimet Hengityssuojaimet
Disposable FFP3-mask consists of a PP substrate to which a laminated microporous polyethlene film is applied (PP/PE). The outer layer of fabric prevents the passage of liquid molecules inwards yet permits the vapour molecules that build up inside to pass through.
  • Adjustable nose bridge
  • Sweat absorbing nose foam (silicone/latex-free)
  • Twin elastic headband
  • Colour coded to indicate performance level
  • With easy flow exhalation valve
  • Can be used in concentrations of contaminats up to 20 times WEL
Sertifikaatit: EN 149:2001+A1:2009