Maximum cut protection nitrile foam gloves, A645
Toimittaja: Portwest
A645E8RS A645E8RM A645E8RL A645E8RXL
PRTWA645E8RSEA 120 EUR
PRTWA645E8RS PRTWA645E8RM PRTWA645E8RL PRTWA645E8RXL
Maximum cut protection nitrile foam gloves, A645
Käsineet
Nitrile foam coating in the gloves provides excellent grip in wet and dry conditions. Bright green liner provides easy identification.
  • Level D cut resistance
  • Green - High risk cut protection
  • Palm dipped to increase dexterity and ventilation
  • Protects against cuts and abrasion
  • Retail bag which aids presentation for retail sales
  • 13 gauge liner for a perfect fit
  • 100% breathable seamless liner
Sertifikaatit: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level A4. EN 388:2003 - 4.5.4.4. EN 388:2016 - 4.X.4.4.D. EN 420. CE Food Safe. EN cut protection levels.