Anti-Listeria Chicken Polyclonal Antibody
Toimittaja: ProSci Inc.
XW-7705
PRSIXW-7705EA 918 EUR
PRSIXW-7705
Anti-Listeria Chicken Polyclonal Antibody
Vasta-aineet
FUNCTION: Produces green light with a wavelength of 562 nM.
CATALYTIC ACTIVITY: Photinus luciferin + O2 + ATP = oxidized Photinus luciferin + CO2 + AMP + diphosphate + light.
SIMILARITY: Belongs to the ATP-dependent AMP-binding enzyme family.
COFACTOR: Magnesium.
SUBCELLULAR LOCATION: Peroxisomal.
Luciferin 4-monooxygenase antibody can be used for the detection of Luciferin 4-monooxygenase by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.