Anti-hemagglutinin Mouse Monoclonal Antibody [clone: 4E10C10 ]
Toimittaja: ProSci Inc.
PM-4669
PRSIPM-4669EA 435 EUR
PRSIPM-4669
Anti-hemagglutinin Mouse Monoclonal Antibody [clone: 4E10C10 ]
Vasta-aineet
Hemagglutinin Monoclonal Antibody: Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals, but it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. HA interacts with host cell surface proteinsMore Product Information