Anti-CD72.1 Mouse Monoclonal Antibody [clone:10.1.D2]

Toimittaja: ProSci Inc.
98-840
PRSI98-840EA 547 EUR
PRSI98-840
Anti-CD72.1 Mouse Monoclonal Antibody [clone:10.1.D2]
Vasta-aineet
Type: Primary
Antigen: CD72.1
Clonality: Monoclonal
Clone: 10.1.D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR