Human recombinant Chymotrypsin-like Elastase 3A (from cells)
Toimittaja: ProSci Inc.
91-740
PRSI91-740EA 884 EUR
PRSI91-740
Human recombinant Chymotrypsin-like Elastase 3A (from cells)
Entsyymit
Chymotrypsin-Like Elastase Family Member 3A (CELA3A) is an enzyme that contains one peptidase S1 domain. ELA3A belongs to the peptidase S1 family of the Elastase subfamily. ELA3A is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein, it has a digestive function in the intestine. ELA3A may also function in the intestinal transport and metabolism of cholesterol. ELA3A is efficient protease with alanine specificity but only little elastolytic activity. ELA3A preferentially cleaves proteins after alanine residues.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.