Human recombinant IL-20 R alpha (from cells)
Toimittaja: ProSci Inc.
91-537
PRSI91-537EA 549 EUR
PRSI91-537
Human recombinant IL-20 R alpha (from cells)
Proteiinit
Interleukin-20 Receptor Subunit alpha (IL20RA) is a single-pass type I membrane protein that is a member of the type II cytokine receptor family. IL20RA is synthetized a 553 amino acid glycoprotein precursor containing a 29 amino acid signal peptide, a 221 amino acid extracellular domain with two fibronectin type-III domains, a 24 amino acid transmembrane region, and a 279 amino acid intracellular domain. IL20RA is widely expressed with highest levels found in skin and testis and high levels in brain. IL20RA forms a heterodimer with IL20RB, and the complex serves as a receptor for IL19, IL20 and IL24. IL20RA also forms a heterodimer with the unique and specific receptor IL10RB and functions as the receptor for IL26.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag