Anti-CD69 Mouse Monoclonal Antibody [clone: FN50] (PE (Phycoerythrin))

Toimittaja: ProSci Inc.
76-816
PRSI76-816EA 311 EUR
PRSI76-816
Anti-CD69 Mouse Monoclonal Antibody [clone: FN50] (PE (Phycoerythrin))
Vasta-aineet
The FN50 monoclonal antibody specifically reacts with human CD69, the 27 to 33 kDa type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR