Anti-XRN2 Rabbit Polyclonal Antibody

Toimittaja: ProSci Inc.
57-673
PRSI57-673EA 620 EUR
PRSI57-673
Anti-XRN2 Rabbit Polyclonal Antibody
Vasta-aineet
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: XRN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 48-76
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR