Anti-NUDT9 Rabbit Polyclonal Antibody

Toimittaja: ProSci Inc.
56-294
PRSI56-294EA 620 EUR
PRSI56-294
Anti-NUDT9 Rabbit Polyclonal Antibody
Vasta-aineet
NUDT9 hydrolyzes ADP-ribose (ADPR) to AMP and ribose 5'-phosphate.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.

Type: Primary
Antigen: NUDT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR