Anti-TNFSF13B Mouse Monoclonal Antibody [clone: C257/1638]
Toimittaja: ProSci Inc.
33-595
PRSI33-595EA 773 EUR
PRSI33-595
Anti-TNFSF13B Mouse Monoclonal Antibody [clone: C257/1638]
Vasta-aineet
BAFF/BLyS/CD257 is a type II transmembrane protein, and a member of the tumor necrosis factor ligand superfamily. It is proteolytically cleaved to form a soluble protein. BAFF, like other TNF ligand family members, forms homo-trimers. The predicted molecular weight of a BAFF monomer is approximately 31 kDa. It is expressed on peripheral blood B and T lymphocytes, monocytes, macrophages, and dendritic cells and is upregulated by IFN-gamma and down-regulated by PMA/ionomycin treatment. BAFF stimulates B and T cell immunity and regulates humoral immunity by binding TACI and BCMA receptors; BAFFMore Product Information