Anti-ICAM3 Mouse Monoclonal Antibody [clone: ICAM3/1019]
Toimittaja: ProSci Inc.
33-402
PRSI33-402EA 773 EUR
PRSI33-402
Anti-ICAM3 Mouse Monoclonal Antibody [clone: ICAM3/1019]
Vasta-aineet
Recognises an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This mAb is excellent for staining of formalin/paraffin tissues.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) PredilutedMore Product Information