Anti-CDH16 Mouse Monoclonal Antibody [clone: CDH16/1071]
Toimittaja: ProSci Inc.
33-309
PRSI33-309EA 773 EUR
PRSI33-309
Anti-CDH16 Mouse Monoclonal Antibody [clone: CDH16/1071]
Vasta-aineet
This mAb recognises a protein of 130 kDa, identified as Ksp-cadherin. Cadherins form a superfamily of related glycoproteins that mediate calcium-dependent cell adhesion and transmit signals from the extracellular matrix to the cytoplasm. Cadherins have been implicated in embryogenesis, tissue morphogenesis, tissue structure maintenance, cell polarization, neoplastic invasiveness and metastasis, and membrane transport. It is suggested that Ksp-cadherin is a marker for terminal differentiation of the basolateral membranes of renal tubular epithelial cells. Within the kidney, Ksp-Cadherin isMore Product Information