Anti-NMNAT1 Rabbit Polyclonal Antibody
Toimittaja: ProSci Inc.
30-467
PRSI30-467EA 601 EUR
PRSI30-467
Anti-NMNAT1 Rabbit Polyclonal Antibody
Vasta-aineet
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca (2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD.The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca (2+)More Product Information