Anti-PCYT2 Rabbit Polyclonal Antibody

Toimittaja: ProSci Inc.
29-889
PRSI29-889EA 601 EUR
PRSI29-889
Anti-PCYT2 Rabbit Polyclonal Antibody
Vasta-aineet
The function remains unknown.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PCYT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR