Anti-MKNK2 Rabbit Polyclonal Antibody

Toimittaja: ProSci Inc.
26-999
PRSI26-999EA 660 EUR
PRSI26-999
Anti-MKNK2 Rabbit Polyclonal Antibody
Vasta-aineet
MKNK2 may play a role in the response to environmental stress and cytokines. It appears to regulate transcription by phosphorylating EIF4E, thus increasing the affinity of this protein for the 7-methylguanosine-containing mRNA cap.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MKNK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR