Anti-INSR Recombinant Antibody [Clone: 83-7]

Toimittaja: ProSci Inc.
24-176
PRSI24-176EA 737 EUR
PRSI24-176
Anti-INSR Recombinant Antibody [Clone: 83-7]
Vasta-aineet
Type: Primary
Antigen: insulin receptor
Clonality: Recombinant
Clone: 83-7
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR