Anti-ZFAND3 Rabbit Polyclonal Antibody

Toimittaja: ProSci Inc.
22-970
PRSI22-970EA 419 EUR
PRSI22-970
Anti-ZFAND3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZFAND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR