Cryopreservation solution, DMSO-free, Cellvation™
Toimittaja: PROTIDE PHARMACEUTICALS
5001
PRPH5001EA 0 EUR
PRPH5001
Cryopreservation solution, DMSO-free, Cellvation™
Soluviljelyyn suplementit ja lisäaineet
A unique cryopreservation solution that does not contain DMSO or foetal bovine serum. Cellvation™ is convenient to use and provides optimal recovery. Cells can be exposed to Cellvation™ for longer periods of time before the freezing process is initiated. Cellvation™ is ready to use and can be used for cells cultured in serum-free medium or with serum-containing medium.
  • Does not contain DMSO
  • Sterile/ready to use
Sertifikaatit: cGMP manufactured.