Elemental analysis standards, sulphur standards, VHG
Toimittaja: LGC Standards PROMOCHEM
VHGSDSL1500100 VHGSDSL200100 VHGSDSL25100 VHGSDSL2P100 VHGSDSL3000100 VHGSDSL3P100 VHGSDSL400100 VHGSDSL4P100 VHGSDSL5000100 VHGSDSL500100 VHGSDSL5P100 VHGSDSL7500100 VHGSDSL5100 VHGSDSL1000100 VHGSDSL300100 VHGSDSL10100 VHGSDSL15100 VHGSDSL20100 VHGSDSL50100 VHGSDSL75100 VHGSDSL1P100 VHGSDSL100100 VHGSDSL750100 VHGSDSLBLK100
PROOVHGSDSL1500100EA 223 EUR
PROOVHGSDSL1500100 PROOVHGSDSL200100 PROOVHGSDSL25100 PROOVHGSDSL2P100 PROOVHGSDSL3000100 PROOVHGSDSL3P100 PROOVHGSDSL400100 PROOVHGSDSL4P100 PROOVHGSDSL5000100 PROOVHGSDSL500100 PROOVHGSDSL5P100 PROOVHGSDSL7500100 PROOVHGSDSL5100 PROOVHGSDSL1000100 PROOVHGSDSL300100 PROOVHGSDSL10100 PROOVHGSDSL15100 PROOVHGSDSL20100 PROOVHGSDSL50100 PROOVHGSDSL75100 PROOVHGSDSL1P100 PROOVHGSDSL100100 PROOVHGSDSL750100 PROOVHGSDSLBLK100
Elemental analysis standards, sulphur standards, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Sulphur Standards
This standard solution is intended for use as a certified reference material and calibration standard for the analysis of sulphur in various petroleum products using x-ray fluorescence spectrometry (XRF), combustion and other techniques for sulphur analysis. It is suitable for use with methods such as ASTM D2622, D4294, D5453, D7039, D7212, D7220 and others.
Sertifikaatit: Traceable to NIST and SRM.