Lutetium standard solution, for ICP, in HNO₃, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PLUN-100 VHG-LLUN-100 VHG-PLUN-500 VHG-TLUN-100 VHG-TLUN-500
PROOVHG-PLUN-100EA 187 EUR
PROOVHG-PLUN-100 PROOVHG-LLUN-100 PROOVHG-PLUN-500 PROOVHG-TLUN-100 PROOVHG-TLUN-500
Lutetium standard solution, for ICP, in HNO₃, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Lutetium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.